Learn More
Bis(maltolato)oxovanadium(IV), TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Bis(maltolato)oxovanadium(IV) |
| CAS | 38213-69-3 |
| Summenformel | C12 H10 O7 V |
| Menge | 250 mg |
| Synonym | Vanadium, bis[3-(hydroxy-κO)-2-methyl-4H-pyran-4-onato-κO4]oxo-, (SP-5-31)-, Vanadium, bis(3-hydroxy-2-methyl-4H-pyran-4-onato-O3,O4)oxo-, (SP-5-31)-, (SP-5-31)-Bis[3-(hydroxy-κO)-2-methyl-4H-pyran-4-onato-κO4]oxovanadium, 4H-Pyran-4-one, 3-hydroxy-2-methyl-, vanadium complex, Bis(maltolato)oxovanadium, Bis(maltolato)oxovanadium(IV) |
| InChI-Formel | InChI=1S/2C6H6O3.O.V/c2*1-4-6(8)5(7)2-3-9-4;;/h2*2-3,8H,1H3;;/q;;;+2/p-2 |
| SMILES | CC1=C(O[V](=O)OC2=C(C)OC=CC2=O)C(=O)C=CO1 |
| IUPAC-Name | Bis[(2-Methyl-4-oxopyran-3-yl)oxy]-oxovanadium |
| Molekulargewicht (g/mol) | 317.15 |
| Formelmasse | 316.9866 |