missing translation for 'onlineSavingsMsg'
Learn More

Bis(p-nitrobenzyl)phosphat, TRC

Product Code. 30340825
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Menge:
100 mg
missing translation for 'unitSize'
100 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30340825 100 mg 100 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30340825 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCB504070100MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Specifications

Chemischer Name oder Material Bis(p-nitrobenzyl) Phosphate
CAS 14390-40-0
Summenformel C14H13N2O8P
Menge 100 mg
Synonym 4-Nitro-benzenemethanol Hydrogen Phosphate, p-Nitro-benzyl Alcohol Hydrogen Phosphate, NSC 401700
InChI-Formel InChI=1S/C14H13N2O8P/c17-15(18)13-5-1-11(2-6-13)9-23-25(21,22)24-10-12-3-7-14(8-4-12)16(19)20/h1-8H,9-10H2,(H,21,22)/p-1
SMILES [O-][N+](=O)c1ccc(COP(=O)([O-])OCc2ccc(cc2)[N+](=O)[O-])cc1
IUPAC-Name Bis[(4-Nitrophenyl)methyl]phosphat
Molekulargewicht (g/mol) 368.24
Formelmasse 367.0331
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.