Learn More
BIVM Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25432-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
BIVM has been identified using an electronic search based on the conservation of short sequence motifs within the variable region of immunoglobulin (Ig) genes. It is tightly linked (41 bp) and in the opposite transcriptional orientation to MGC5302 (also known as KDEL1 and EP58) in human. BIVM has two isoforms with MW 57 kDa and 32 kDa.Spezifikation
BIVM | |
Polyclonal | |
Unconjugated | |
BIVM | |
BIVM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54841 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86UB2 | |
BIVM | |
BIVM Fusion Protein Ag22087 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |