missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15964-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is a member of the BCL2/adenovirus E1B 19 kd-interacting protein family. It interacts with the E1B 19 kDa protein, which protects cells from virally-induced cell death. The encoded protein also interacts with E1B 19 kDa-like sequences of BCL2, another apoptotic protector. In addition, this protein is involved in vesicle transport into the endoplasmic reticulum. Alternative splicing of this gene results in four protein products with identical N- and C-termini.Spezifikation
BNIP1 | |
Polyclonal | |
Unconjugated | |
BNIP1 | |
BNIP1, NIP1, SEC20, SEC20L, TRG 8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140932, 224630, 662 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q12981, Q6QD59, Q8VHI8 | |
BNIP1 | |
BNIP1 Fusion Protein Ag8737 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |