missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26080-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene is located within a region of a segmental duplication on chromosome 16 and is identical to BOLA2B (bolA family member 2B). The product of this gene belongs to a family of proteins that are widely conserved and may be involved in iron maturation. Related pseudogenes are found multiple different chromosomes. Alternative splicing results in multiple transcript variants. Transcripts initiating at this locus may extend into downstream SMG1 pseudogene 6 (SMG1P6) and encode fusion proteins with a C-terminus related to SMG1 phosphatidylinositol 3-kinase-related kinase. A readthrough locus is represented with GeneID:107282092.Spezifikation
BOLA2 | |
Polyclonal | |
Unconjugated | |
BOLA2 | |
bolA homolog 2 (E. coli), BolA like protein 2, BOLA2, BOLA2A, BOLA2B, My016 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
552900 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H3K6 | |
BOLA2 | |
BOLA2 Fusion Protein Ag23260 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |