missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BORIS Mouse anti-Human, Mouse, Clone: 2B12E2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 60079-1-IG-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
CTCFL is a paralog of CTCF and appears to be expressed primarily in the cytoplasm of spermatocytes. CTCFL is normally expressed in a mutually exclusive pattern that correlates with resetting of methylation marks during male germ cell differentiation.Spezifikation
BORIS | |
Monoclonal | |
0.71 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A2APF3, Q8NI51 | |
Ctcfl | |
BORIS Fusion Protein Ag1003 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Western Blot | |
2B12E2 | |
Unconjugated | |
CTCFL | |
BORIS, Cancer/testis antigen 27, CCCTC binding factor, CT27, CTCF like protein, CTCF paralog, CTCF T, CTCFL, Zinc finger protein CTCF T | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
140690, 664799 | |
-20°C | |
Liquid |