missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromine, Reagent grade, Honeywell
Click to view available options
Menge:
100 mL
500 mL
1 L
Spezifikation
Chemischer Name oder Material | Bromine |
Menge | 100 mL |
UN-Nummer | 1744 |
Formelmasse | 159.81g/mol |
Güte | Reagent |
Dampfdruck | 671mmHg (55°C), 175mmHg (20°C) |
Dichte | 3.119g/mL at 25°C; Vapor Density: 7.14 (vs air) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'