missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Bromodeoxyuridine (BrdU) Ab-3 Mouse Monoclonal Antibody, 200μg/mL, biotinylated, BSA and azide
Detect S-phase cells with Epredia™ Bromodeoxyuridine (BrdU) Ab-3, Mouse Monoclonal Antibody, which provides accurate, reproducible results.
289.92 €
Spezifikation
Antigen | Bromodeoxyuridine (BrdU) Ab-3 |
---|---|
Klon | BRD.3 |
Konzentration | 40μg/mL |
Anwendungen | Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Klassifikation | Monoclonal |
Produktcode | Marke | Form | Menge | Preis | Menge & Verfügbarkeit | ||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Form | Menge | Preis | Menge & Verfügbarkeit | ||||
13462679
|
Epredia™
MS-1058-P1 |
Concentrated | 500 μL |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
|||||
Beschreibung
BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.Host Species: Mouse
Clone: BRD.3
Isotype: IgG1
Species Reactivity: All species.
Immunogen: Bromodeoxyuridine (BrdU) conjugated to BSA
Positive Control: Cultured cells grown in presence of BrdU or tissues from experimental animals injected with BrdU
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
- Flow Cytometry
Note:
USA: RUO; Int'l: RUO
Spezifikation
Bromodeoxyuridine (BrdU) Ab-3 | |
40μg/mL | |
Monoclonal | |
Mouse | |
Epitope Tags | |
Bromodeoxyuridine (BrdU) conjugated to BSA | |
Primary |
BRD.3 | |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Concentrated | |
RUO | |
All species | |
IgG1 |