Learn More
Bromoethane, ≥99%, extra pure, Thermo Scientific Chemicals
Product Code. 10264701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10264701 100 mL Glass bottle 100mL
10173381 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product contains an ozone depleters according to Regulation (EC) No 1005/2009, all customers must complete a declaration before delivery.
Product Code. 10264701 Supplier Thermo Scientific Acros Supplier No. 330351000
 Restricted Item
Lager vorrätig
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product contains an ozone depleters according to Regulation (EC) No 1005/2009, all customers must complete a declaration before delivery.
CAS: 74-96-4 | Molecular Formula: C2H5Br | Molecular Weight: 108.966 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 74-96-4
Molecular Formula C2H5Br
Molecular Weight (g/mol) 108.966
MDL Number MFCD00000232
InChI Key RDHPKYGYEGBMSE-UHFFFAOYSA-N
Synonym ethyl bromide, ethane, bromo, 1-bromoethane, monobromoethane, bromic ether, hydrobromic ether, bromure d'ethyle, ethylbromide, etylu bromek, halon 2001
PubChem CID 6332
IUPAC Name bromoethane
SMILES CCBr
Specifications
Acidity 0.0005 % max. (as HBr)
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula C2H5Br
Refractive Index 1.4230 to 1.4250 (20°C, 589nm)
Quantity 100 mL
Formula Weight 108.97
Percent Purity ≥99.0% (GC)
Grade Extra Pure
Physical Form Crystals or Powder
Chemical Name or Material Bromoethane, 99+%