Learn More
BTD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562581
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NLDIYEQQVM TAAQKDVQII VFPEDGIHGF NFTRTSIYPF LDFMPSPQVV RWNPCLEPHR FNDTEVLQRL SCMAIRGDMF LVA Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 86%.
Biotinidase functions to recycle biotin in the body by cleaving biocytin (biotin-epsilon-lysine), a normal product of carboxylase degradation, resulting in regeneration of free biotin. Biotinidase has also been shown to have biotinyl-transferase activity. Defects in the biotinidase gene cause multiple carboxylase deficiency.Spezifikation
BTD | |
Polyclonal | |
Unconjugated | |
Btd | |
biotinase; Biotinidase; Btd; btd {ECO:0000312; EC 3.5.1.12; EMBL:AAH90017.1} | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
686 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P43251 | |
Btd | |
Recombinant protein corresponding to Human BTD | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only