missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BTN2A1 Recombinant Protein
Marke: Abnova™ H00011120-P01.10ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH16661 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
62.48 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BTN2A1 | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
11120 | |
BTN2A1 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
BK14H9.1/BT2.1/BTF1/DJ3E1.1/FLJ36567 | |
BTN2A1 | |
Wheat Germ (in vitro) | |
GST |