missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 10, Mettler Toledo™
Click to view available options
Quantity: 6 x 250mL
Spezifikation
Farbe | Yellow |
pH | 10 |
Physikalische Form | Liquid |
Menge | 250 mL |
Lösungstyp | pH Buffer |
Zusammensetzung | Kaliumhexacyanoferrat(II), Kaliumhexacyanoferrat(III), 0.01 to 0.1% Sodium hydroxide; caustic soda |
Geruch | None |
Verpackung | 6 Pk. |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'