missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solutions, Merck Chemicals™
Click to view available options
Menge:
30 x 30 mL
Spezifikation
Farbe | Colorless |
Physikalische Form | Liquid |
Menge | 30 x 30 mL |
Geruch | Odorless |
Löslichkeitsinformationen | Soluble in water |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'