Learn More
Fisherbrand™ Halogenlampe 12V, 10W, G4, für diaskop. Licht
Illuminate Inverso TC100 Microscopes with this light bulb. Fisherbrand™ Bulbs for Diascopic Light are ergonomically designed for everyday use.
Marke: Fisherbrand™
Weitere Details : Netto-Gewicht : 1.00000kg
Beschreibung
- G4 Halogen Bulb for diascopic light
- Accessory for microscopes: FB69155 and FB69156
- Available in packs of ten