missing translation for 'onlineSavingsMsg'
Learn More

Merck Chemicals Butylated Hydroxytoluene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Product Code. 17819851
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17819851 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17819851 Supplier Merck Chemicals Supplier No. PHR11171G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Butylated hydroxytoluene (BHT) is a phenolic antioxidant used as a food additive. It is frequently added to dried cereals, cooking oils and animal foods. BHT is also known to enhance the stability of pharmaceuticals, fat-soluble vitamins and cosmetics . Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 128-37-0
Ignition Point 878°F
Vapor Pressure <0.01 mmHg (20°C)
Vapor Density 7.6 (vs. air)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula [(CH3)3 C]2C6H2(CH3)OH
MDL Number MFCD00011644
Melting Point 69°C to 73°C
Boiling Point 265°C
Synonym 2,6-Di-tert-butyl-4-methylphenol; 2,6-Di-tert-butyl-p-cresol; BHT; Butylated hydroxytoluene; Butylhydroxytoluene; DBPC
Type Certified Reference Material
Quantity 1 g
Formula Weight 220.35
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.