Learn More
BVES Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553235
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LNDPTLNDKK AKKLEHQLSL CTQISMLEMR NSIASSSDSD DGLHQFLRGT SSMSSLHVSS PHQRASAKMK PIEEGAEDDD DVFEPASPNT LKVHQLP Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 25%.
BVES Polyclonal antibody specifically detects BVES in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
BVES | |
Polyclonal | |
Unconjugated | |
BVES | |
blood vessel epicardial substance; bves; hBVES; LGMD2X; mBVES; MGC42413; POP 1; POP1; POPDC 1; Popdc1; popdc1 protein; popeye 1; popeye domain containing 1; popeye domain-containing 1; popeye domain-containing protein 1; Popeye protein 1; zgc:86887 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11149 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NE79 | |
BVES | |
Recombinant protein corresponding to Human BVES | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only