Learn More
C/EBP gamma Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552895
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GVNGISVIHT QAHASGLQQV PQLVPAGPGG GGKAVAPSKQ SKKSSPMDRN SDEYRQRRER NNMAVKKSRL KSKQKAQDTL QRVNQLKEEN ERLEAKIKLL TKELSVLKDL FLEHAHNLAD NVQSISTENT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 98%.
C/EBP gamma Polyclonal antibody specifically detects C/EBP gamma in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C/EBP gamma | |
Polyclonal | |
Unconjugated | |
CEBPG | |
C EBP; C/EBP; c/EBP gamma; C/EBP[g]; C/EBPgamma; CCAAT/enhancer binding protein (C/EBP), gamma; CCAAT/enhancer binding protein ,gamma; CCAAT/enhancer binding protein gamma; CCAAT/enhancer-binding protein gamma; CEBP gamma; CEBPG; CEBPRNA; G-CSF promoter element 1 binding protein; GPE1-binding protein; Gpe1bp; GPE1-BP; GPE1-BP (C/EBP gamma); Granulocyte colony-stimulating factor promoter element 1-binding protein; homolog to a human CCAAT/enhancer binding protein - gamma; Ig/EBP; IG/EBP-1; immunoglobulin enhancer-binding protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1054 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P53567 | |
CEBPG | |
Recombinant protein corresponding to Human C/EBP gamma | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only