Learn More
C10orf118 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA553195
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LRSSTFPESA NEKTYSESPY DTDCTKKFIS KIKSVSASED LLEEIESELL STEFAEHRVP NGMNKGEHAL VLFEKCVQDK YLQQEHIIKK LIKENKKHQE LFVDICSEKD NLREELKKRT ETEK Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.
C10orf118 Polyclonal antibody specifically detects C10orf118 in Human samples. It is validated for Immunohistochemistry.Spezifikation
C10orf118 | |
Polyclonal | |
Unconjugated | |
CCDC186 | |
1810028B20Rik; A630007B06Rik; AI159700; C10orf118; CCDC186; coiled-coil domain containing 186; coiled-coil domain-containing protein 186; CTCL tumor antigen HD-CL-01/L14-2; CTCL tumor antigen L14-2; FLJ10188; FLJ35301; oocyte-testis gene 1 protein; Otg1; RGD1307158 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55088 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z3E2 | |
CCDC186 | |
Recombinant protein corresponding to Human C10orf118 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only