missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ C10orf7 293T Cell Transient Overexpression Lysate (Denatured)
Specifications
Specifications
| Zelllinie | 293T |
| Zur Verwendung mit (Anwendung) | Western Blot |
| Beschreibung | C10orf7 293T Cell Transient Overexpression Lysate(Denatured) |
| Gen-Zugriffsnummer | BC001600 |
| Molekulargewicht | Theoretical M.W. 39.1kDa |
| Plasmid | pCMV-C10orf7 full-length |
| Qualitätskontrollen | Transient overexpression cell lysate was tested with anti-C10orf7 antibody (Mfr. No. H00008872-D01) by Western Blot. |
| Menge | 100 μl |
| Lagerungspuffer | 1X Probenpuffer (50 mM Tris-HCl, 2 % SDS, 10 % Glycerin, 300 mM 2-Mercaptoethanol, 0,01 % Bromophenolblau) |
| Wirtsspezies | Human |
| Show More |
Safety and Handling
Product Identifier
- 100UL C10orf7 293T Cell Transient Overexpression L
- Achtung
- Sensibilisierung der Haut Kategorie 1
- H317-Kann allergische Hautreaktionen verursachen.
- P261-Einatmen von Staub/Rauch/Gas/Nebel/Dampf/Aerosol vermei den.
- P272-Kontaminierte Arbeitskleidung nicht außerhalb des Arbeitsplatzes tragen.
- P280-Schutzhandschuhe/Schutzkleidung/Augenschutz/Gesichtsschutz tragen.
- MIXTURE LIST-Contain: Mercaptoethanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?