Learn More
C10orf76 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562812
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ELPLDADVQT SNLLITFLKY SSIVMQDTKD EHRLHSGKLC LIILTCIAED QYANAFLHDD NMNFRVNLHR MPMRHRKKAA DKNLPCRP Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
C10orf76 Polyclonal antibody specifically detects C10orf76 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
C10orf76 | |
Polyclonal | |
Unconjugated | |
C10orf76 | |
Armadillo-like helical domain-containing protein 3; ARMH3; C10orf76; chromosome 10 open reading frame 76; UPF0668 protein C10orf76 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79591 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T2E6 | |
C10orf76 | |
Recombinant protein corresponding to Human C10orf76 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only