Learn More
C10orf88 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559380
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PPAPGQDLVI LKRNHNNKDE NPCFLYLRCG PDGGEEIASI GILSSARNME VYLGEEYCGT SRGKNVCTVL D Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.
C10orf88 Polyclonal antibody specifically detects C10orf88 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
C10orf88 | |
Polyclonal | |
Unconjugated | |
C10orf88 | |
C10orf88; chromosome 10 open reading frame 88; uncharacterized protein C10orf88 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80007 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H8K7 | |
C10orf88 | |
Recombinant protein corresponding to Human C10orf88 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only