Learn More
C11orf52 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558367
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MGNRVCCGGS WSCPSTFQKK KKTGSQTRRT LKPQPQQLQQ NLPKGHETTG HTYERVLQQQ GS Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 65%.
C11orf52 Polyclonal antibody specifically detects C11orf52 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C11orf52 | |
Polyclonal | |
Unconjugated | |
C11orf52 | |
C11orf52; chromosome 11 open reading frame 52; uncharacterized protein C11orf52 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91894 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96A22 | |
C11orf52 | |
Recombinant protein corresponding to Human C11orf52 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only