Learn More
C12orf56 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563380
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RSLKESILRD QQESSTPSKD STLCPRPGLK KLSLHGQGAF RPLPSPSRRS SQSAPTTGKA VSEPSCTTNT KEPQGLPDHN SIS Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 50%.
The function of this protein remains unknown.Spezifikation
C12orf56 | |
Polyclonal | |
Unconjugated | |
C12orf56 | |
C12orf56; chromosome 12 open reading frame 56; uncharacterized protein C12orf56 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
115749 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IXR9 | |
C12orf56 | |
Recombinant protein corresponding to Human C12orf56 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only