Learn More
C13orf27 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562802
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LGGRSMGSRA AASVMCHIEP DDGDDFVRGL ICISYPLHHP KQQHKLRDED LFRLKEPVLF VSGSADEMCE KNLLEKVAQK MQAP Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
C13orf27 Polyclonal antibody specifically detects C13orf27 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
C13orf27 | |
Polyclonal | |
Unconjugated | |
TEX30 | |
1700029F09Rik; 3110030D08Rik; C13orf27; testis expressed 30; Testis-expressed protein 30; testis-expressed sequence 30 protein; TEX30 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
93081 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JUR7 | |
TEX30 | |
Recombinant protein corresponding to Human C13orf27 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only