Learn More
C13orf7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557060
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RSENEKKSEC FSSPKTGFWD CCSTSYAQNL DFESSEGNTI ANSVGEISSK LSEKSGLCLS KRLNSIRSFE MNRTRTSSEA SMDAAYLDKI SELDSMMS Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 84%.
C13orf7 Polyclonal antibody specifically detects C13orf7 in Human samples. It is validated for Immunohistochemistry.Spezifikation
C13orf7 | |
Polyclonal | |
Unconjugated | |
RNF219 | |
2610206B13Rik; 2810449K13Rik; AI451544; C13orf7; FLJ13449; H12C13ORF7; Obi1; ORC ubiquitin ligase 1; RGD1564706; RING finger protein 219; RING finger protein 219 pseudogene; RNF219 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79596 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5W0B1 | |
RNF219 | |
Recombinant protein corresponding to Human C13orf7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only