Learn More
C17orf85 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553033
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EYPPAHIEWL DDTSCNVVWL DEMTATRALI NMSSLPAQDK IRSRDASEDK SAEKRKKDKQ EDSSDDDEAE EGEVEDENSS DVELDTLSQV EEESLLRNDL RPANKLAKGN Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
C17orf85 Polyclonal antibody specifically detects C17orf85 in Human samples. It is validated for Immunohistochemistry.Spezifikation
C17orf85 | |
Polyclonal | |
Unconjugated | |
NCBP3 | |
1200014J11Rik; C11H17orf85; C130061O14Rik; C17orf85; C19H17orf85; C78393; C9H17orf85; CTD-3195I5.1; CTD-3195I5.5; ELG; ELG protein; HSA277841; Ncbp3; nuclear cap binding subunit 3; nuclear cap-binding protein subunit 3; Protein ELG; RGD1308139; RIKEN cDNA 1200014J11 gene; similar to RIKEN cDNA 1200014J11; uncharacterized protein C17orf85 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55421 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53F19 | |
NCBP3 | |
Recombinant protein corresponding to Human C17orf85 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only