Learn More
C17orf89 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke:  Thermo Scientific PA563474
Weitere Details : Netto-Gewicht : 0.02000kg
Artikelnummer. 15721252
  • 514.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
Immunogen sequence: LSKDFCAREF EALRSCFAAA AKKTL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
C17orf89 Polyclonal antibody specifically detects C17orf89 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Spezifikation
Spezifikation
C17orf89
Polyclonal
Unconjugated
NDUFAF8
1810043H04Rik; C17orf89; C9H17orf89; chromosome 17 open reading frame 89; hypothetical protein LOC690871; LOC690871; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 8; NADH:ubiquinone oxidoreductase complex assembly factor 8; Ndufaf8; RIKEN cDNA 1810043H04 gene; uncharacterized protein C17orf89; uncharacterized protein C17orf89 homolog
Rabbit
Antigen affinity chromatography
RUO
284184
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
A1L188
NDUFAF8
Recombinant protein corresponding to Human C17orf89