Learn More
C18orf21 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA564323
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: TCNRTVKHHG KSRSFVSTLK SNPATPTSKL SLKTPERRTA NPNHDMSGSK G Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 56%.
C18orf21 Polyclonal antibody specifically detects C18orf21 in Human samples. It is validated for Immunohistochemistry.Spezifikation
C18orf21 | |
Polyclonal | |
Unconjugated | |
C18orf21 | |
C18orf21; chromosome 18 open reading frame 21; HBV XAg-transactivated protein 13; HBV X-transactivated gene 13 protein; HBV X-transactivated protein 13; HsT3108; PNAS-124; PNAS-131; UPF0711 protein C18orf21; XTP13 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83608 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q32NC0 | |
C18orf21 | |
Recombinant protein corresponding to Human C18orf21 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only