Learn More
C19orf68 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563923
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RRLLSYCKGR DHGVLDALHV LEGLFRTDPE AKVKLVFVED QAVVETVFFL TSRTRALLRR FPRMLLVDRL Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.
C19orf68 Polyclonal antibody specifically detects C19orf68 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
C19orf68 | |
Polyclonal | |
Unconjugated | |
ZSWIM9 | |
C19orf68; chromosome 19 open reading frame 68; uncharacterized protein C19orf68; Uncharacterized protein ZSWIM9; ZSWIM9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
374920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86XI8 | |
ZSWIM9 | |
Recombinant protein corresponding to Human C19orf68 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only