missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf151 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563797
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RMWPLAFGSG MGLGMAYSNC QHDFQAPYLL HGKYVKEQE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 92%.
C1orf151 Polyclonal antibody specifically detects C1orf151 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
C1orf151 | |
Polyclonal | |
Unconjugated | |
MINOS1 | |
2310028O11Rik; C1orf151; MIC10; MICOS complex subunit MIC10; MICOS10; MINOS1; MIO10; mitochondrial contact site and cristae organizing system subunit 10; mitochondrial inner membrane organizing system 1; mitochondrial inner membrane organizing system protein 1; novel DUF543 domain containing protein; RGD1560187; RP5-1056L3.2; UPF0327 protein C1orf151; UPF0327 protein C1orf151 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
440574 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TGZ0 | |
MINOS1 | |
Recombinant protein corresponding to Human C1orf151 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only