Learn More
C1orf210 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553088
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RYHASYRHRP LPETGRGGRP QVAEDEDDDG FIEDNYIQPG TGELGTEGSR DHF Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.
C1orf210 Polyclonal antibody specifically detects C1orf210 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
C1orf210 | |
Polyclonal | |
Unconjugated | |
C1orf210 | |
C1orf210; chromosome 1 open reading frame 210; TEMP; Type III endosome membrane protein TEMP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
149466 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVY1 | |
C1orf210 | |
Recombinant protein corresponding to Human C1orf210 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only