missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf86 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26138-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
C1orf86 is a component of the Fanconi anemia (FA) complex required to recruit the FA complex to DNA interstrand cross-links (ICLs) and promote ICLs repair. Following DNA damage recognizes and binds ′Lys-63′-linked ubiquitin generated by RNF8 at ICLs and recruits other components of the FA complex. It promotes translesion synthesis via interaction with REV1.Spezifikation
C1orf86 | |
Polyclonal | |
Unconjugated | |
FAAP20 | |
C1orf86, FAAP20, FP7162 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
199990 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6NZ36 | |
FAAP20 | |
C1orf86 Fusion Protein Ag23534 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |