Learn More
Epredia™ C1q Complement Antibody
Perform rapid and inexpensive assessment of tissues by immunofluorescence staining with Epredia™ C1q Complement, FITC Labeled Antibody.
Marke: Epredia™ RB-1926-R2
Weitere Details : Netto-Gewicht : 0.00400kg
Beschreibung
Host Species: Rabbit
Species Reactivity: Reacts with human C1q complement.
Immunogen: C1q component isolated from human serum
Positive Control: Skin or kidney fixed and processed in the same manner as the test specimen
Cellular Localization: Diseased skin and kidney tissue (e.g., mesangial proliferative glomerulonephritis)
Recommended for:
- Immunofluorescence
Spezifikation
C1q Complement | |
Polyclonal | |
FITC | |
C1q component isolated from human serum | |
IVD | |
Primary | |
Prediluted |
Immunofluorescence | |
Polyclonal | |
Rabbit | |
2 mL | |
Cancer and Tumor Biology | |
Human |