missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550013
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total C1QB protein.
C1QB Polyclonal antibody specifically detects C1QB in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
C1QB | |
Polyclonal | |
Unconjugated | |
C1QB | |
C1QB; C1QTNF5; complement C1q B chain; complement C1q chain B; complement C1q subcomponent subunit B; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component C1q, B chain; complement subcomponent C1q chain B; CTRP5; DKFZP586B0621; LORD | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
713 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P02746 | |
C1QB | |
Synthesized peptide derived from internal of human C1QB. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |