Learn More
C20orf12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563734
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EKMSDHKPLL TAISPGRGYW RRQLDHISAH LRCYAQNNPE FRALIAEPRM GKLISATVHE DGCEVSIRLN YSQVSNKNLY LNKAVNFSDH LLSSAA Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
C20orf12 Polyclonal antibody specifically detects C20orf12 in Human samples. It is validated for Immunohistochemistry.Spezifikation
C20orf12 | |
Polyclonal | |
Unconjugated | |
DZANK1 | |
ANKRD64; ankyrin repeat domain 64; ankyrin repeat-containing protein C20orf12; bA189K21.1; bA189K21.8; C20orf12; C20orf84; dJ568F9.2; double zinc ribbon and ankyrin repeat domains 1; double zinc ribbon and ankyrin repeat-containing protein 1; DZANK1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55184 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NVP4 | |
DZANK1 | |
Recombinant protein corresponding to Human C20orf12 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only