Learn More
C22orf29 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551574
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PRSLPAAMAT PAVSGSNSVS RSALFEQQLT KESTPGPKEP PVLPSSTCSS KPGPVEPASS QPEEAAPTPV PRLSESANPP AQRPDPAHPG GPKPQKTEEE VLETEGDQEV SLGTPQEVVE APETPGEPPL SP Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 32%.
C22orf29 Polyclonal antibody specifically detects C22orf29 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
C22orf29 | |
Polyclonal | |
Unconjugated | |
RTL10 | |
BH3-only protein; BOP; C22orf29; chromosome 22 open reading frame 29; protein Bop; Retrotransposon Gag-like protein 10; RTL10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79680 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L3V2 | |
RTL10 | |
Recombinant protein corresponding to Human C22orf29 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only