missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C22orf41 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA551209
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total C22orf41 protein.
C22orf41 Polyclonal antibody specifically detects C22orf41 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
C22orf41 | |
Polyclonal | |
Unconjugated | |
SYCE3 | |
C22orf41; SYCE3; Synaptonemal complex central element protein 3; testis highly expressed gene 2 protein; testis highly expressed protein 2; THEG2; THEG-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
644186 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
A1L190 | |
SYCE3 | |
Full length fusion protein of human SYCE3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |