Learn More
C2CD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558437
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ELMVKSSFLS SPERAVNPHL PRQGSPSQSL VACECEASKA RVGGESASAN PQPIPCPTLS GAQQSSTFVG WSSPQTDQNK EPKS Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 39%.
C2CD3 Polyclonal antibody specifically detects C2CD3 in Human samples. It is validated for Immunohistochemistry.Spezifikation
C2CD3 | |
Polyclonal | |
Unconjugated | |
C2CD3 | |
AU020772; C2 calcium dependent domain containing 3; C2 calcium-dependent domain containing 3; C2 domain-containing protein 3; C2CD3; Hty; OFD14; Protein hearty; RGD1307366 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26005 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q4AC94 | |
C2CD3 | |
Recombinant protein corresponding to Human C2CD3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only