Learn More
C2orf42 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556874
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: HKSNASKDET AQRCIHFFAC ICAFASDETL AQEFSDFLNF DSSGLKEIIV PQLGCHSEST VSACESTASK SKKRRKDEVS G Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.
C2orf42 is a protein coding gene.Spezifikation
C2orf42 | |
Polyclonal | |
Unconjugated | |
C2orf42 | |
C2orf42; chromosome 2 open reading frame 42; uncharacterized protein C2orf42 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54980 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NWW7 | |
C2orf42 | |
Recombinant protein corresponding to Human C2orf42 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only