missing translation for 'onlineSavingsMsg'
Learn More

C2orf74 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marque:  Thermo Scientific PA555429

Informations supplémentaires : Netto-Gewicht : 0.02000kg

Code produit. 15733351

  • 514.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Description

Description

Immunogen sequence: DANGGVDCAA AKVVTSNPED HERILMQVMN LNVPMRPGIL VQRQSKEVLA TPLENRRDME AEEENQINEK QEPENAGETG QEEDDGLQKI HTSVTRTPSV VESQKRPLKG VTFSREVIVV DLGNEYPTPR SYT Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 47%.

C2orf74 Polyclonal antibody specifically detects C2orf74 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Spécification

Spécification

C2orf74
Polyclonal
Unconjugated
C2orf74
C2orf74; chromosome 2 open reading frame 74; uncharacterized protein C2orf74
Rabbit
Antigen affinity chromatography
RUO
339804
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
A8MZ97
C2orf74
Recombinant protein corresponding to Human C2orf74
100 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

Research Use Only