Learn More
C2orf91 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA560010
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ALHCPFDFPQ APLRGRHTLS QVPNKGHEKA SAVQLPEKQG TDQSRRGPTS AVTKA Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 33%.
C2orf91 Polyclonal antibody specifically detects C2orf91 in Human samples. It is validated for Immunohistochemistry.Spezifikation
C2orf91 | |
Polyclonal | |
Unconjugated | |
C2orf91 | |
C2orf91; chromosome 2 open reading frame 91; uncharacterized protein C2orf91 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
400950 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZV80 | |
C2orf91 | |
Recombinant protein corresponding to Human C2orf91 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only