missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3a Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550364
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total C3a Receptor protein.
C3a Receptor Polyclonal antibody specifically detects C3a Receptor in Human samples. It is validated for Western Blot.Spezifikation
C3a Receptor | |
Polyclonal | |
Unconjugated | |
C3ar1 | |
anaphylatoxin C3a receptor; AZ3B; C3a anaphylatoxin chemotactic receptor; C3a receptor; C3AR; C3a-R; C3AR1; C3r1; complement component 3 receptor 1; complement component 3a receptor 1; HNFAG09 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
719 | |
-20°C | |
Liquid |
Western Blot | |
1.0 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q16581 | |
C3ar1 | |
Fusion protein corresponding to a region derived from internal residues of human complement component 3a receptor 1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |