Learn More
C3orf1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562760
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: YSGETVQERK QKDRKALHEL KLEEWKGRLQ VTEHLPEKIE SSLQEDEPEN DAKKIEALLN LPRNPSVIDK QDK Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 60%.
C3orf1 Polyclonal antibody specifically detects C3orf1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C3orf1 | |
Polyclonal | |
Unconjugated | |
TIMMDC1 | |
2810021C21Rik; 4930455C21Rik; AV135763; C3orf1; Complex I assembly factor TIMMDC1, mitochondrial; M5-14 protein; Protein M5-14; TIMM domain containing-protein 1; Timmdc1; translocase of inner mitochondrial membrane domain containing 1; translocase of inner mitochondrial membrane domain-containing protein 1; transmembrane protein C3orf1; transmembrane protein C3orf1 homolog; UNQ247/PRO284 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51300 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NPL8 | |
TIMMDC1 | |
Recombinant protein corresponding to Human C3orf1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only