Learn More
C3orf31 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557839
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MALQTLQSSW VTFRKILSHF PEELSLAFVY GSGVYRQAGP SSDQKNAMLD FVFTVDDPVA WHSKNLKKNW SHYSFL Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 76%.
C3orf31 may be involved in the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.Spezifikation
C3orf31 | |
Polyclonal | |
Unconjugated | |
TAMM41 | |
1500001M20Rik; C20H3orf31; C22H3orf31; C3orf31; CDP-DAG synthase; CDP-diacylglycerol synthase; Mitochondrial translocator assembly and maintenance protein 41 homolog; MMP37-like protein, mitochondrial; phosphatidate cytidylyltransferase, mitochondrial; RAM41; TAM41; TAM41 mitochondrial translocator assembly and maintenance homolog; TAM41, mitochondrial translocator assembly and maintenance protein, homolog; TAM41, mitochondrial translocator assembly and maintenance protein, homolog (S. cerevisiae); TAMM41; Unknown (protein for MGC:134188) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
132001 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BW9 | |
TAMM41 | |
Recombinant protein corresponding to Human C3orf31 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only