Learn More
C4orf33 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563930
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: TKWEGKAYLP WSYFPPNVTK FNSFAIHGSK DKRSYEALYP VPQHELQQGQ KPDFHCLEYF KSFNFNTLLG EEWKQPESDL WLI Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
C4orf33 Polyclonal antibody specifically detects C4orf33 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
C4orf33 | |
Polyclonal | |
Unconjugated | |
C4orf33 | |
C4orf33; chromosome 4 open reading frame 33; UPF0462 protein C4orf33 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
132321 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N1A6 | |
C4orf33 | |
Recombinant protein corresponding to Human C4orf33 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only