Learn More
C6orf62 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556504
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KQFLHVLSRK DKTGIVVNNP NQSVFLFIDR QHLQTPKNKA TIFKLCSICL YLPQEQLTHW AVGTIEDHLR PYMPE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
C6orf62 Polyclonal antibody specifically detects C6orf62 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C6orf62 | |
Polyclonal | |
Unconjugated | |
C6orf62 | |
8030460C05Rik; BC005537; C6orf62; C78201; cDNA sequence BC005537; cf062; chromosome 6 open reading frame 62; dJ30M3.2; HBV XAg-transactivated protein 12; HBV X-transactivated gene 12 protein; HBV X-transactivated protein 12; Nbla00237; Uncharacterized protein C6orf62; uncharacterized protein C6orf62 homolog; XTP12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
81688 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9GZU0 | |
C6orf62 | |
Recombinant protein corresponding to Human C6orf62 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only