Learn More
C7orf61 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA563854
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: QVVETLRLVE LPKEAKPKLG ESPELADPCV LAKTTEETEV ELGQQGQSLL QLPRTAVKSV Highest antigen sequence identity to the following orthologs: Mouse - 32%, Rat - 31%.
C7orf61 Polyclonal antibody specifically detects C7orf61 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
C7orf61 | |
Polyclonal | |
Unconjugated | |
C7orf61 | |
C7orf61; chromosome 7 open reading frame 61; uncharacterized protein C7orf61 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
402573 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZ16 | |
C7orf61 | |
Recombinant protein corresponding to Human C7orf61 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only