Learn More
C8G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561200
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: STIQPKANFD AQQFAGTWLL VAVGSACRFL QEQGHRAEAT TLHVAPQGTA MAVSTFRKLD GICWQVRQLY GDTGVLGRFL LQARGARGAV HVVVAET Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 78%.
C8G Polyclonal antibody specifically detects C8G in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
C8G | |
Polyclonal | |
Unconjugated | |
C8G | |
C8C; C8G; complement component 8, gamma polypeptide; complement component C8 gamma chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
733 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P07360 | |
C8G | |
Recombinant protein corresponding to Human C8G | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only