Learn More
C8orf48 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555618
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DFLTRIGEAH QDFPRLSDDP RIIWKRLTEK SHIRYSGFER SETEQKMQRD GNSACHLPFS LPFLKRLTLI KPELVIVNDN V Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 26%.
C8orf48 Polyclonal antibody specifically detects C8orf48 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
C8orf48 | |
Polyclonal | |
Unconjugated | |
C8orf48 | |
C8orf48; chromosome 8 open reading frame 48; uncharacterized protein C8orf48 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
157773 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LL4 | |
C8orf48 | |
Recombinant protein corresponding to Human C8orf48 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only