Learn More
C9orf117 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554508
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: DNQALKSQRD QLSLQLEQQQ VDLQRLQQEL ANEQKVRASL EAALVQATSF LQNILQMHRD EEDSDVDVTF QPWHKEMLQQ LLVMLSST Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 55%.
C9orf117 Polyclonal antibody specifically detects C9orf117 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
C9orf117 | |
Polyclonal | |
Unconjugated | |
CFAP157 | |
1700019L03Rik; C25H9orf117; C9H9orf117; C9orf117; Cfap157; cilia and flagella associated protein 157; cilia- and flagella-associated protein 157; LOC499781; LOW QUALITY PROTEIN: uncharacterized protein C9orf117 homolog; RIKEN cDNA 1700019L03 gene; similar to CG17122-PA; uncharacterized protein C9orf117; uncharacterized protein C9orf117 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
286207 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JU67 | |
CFAP157 | |
Recombinant protein corresponding to Human C9orf117 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only